Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenylacetonitrile, 98+%

Catalog No. AAA1317609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1317609 10 g
AAA1317618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1317609 Supplier Thermo Scientific Chemicals Supplier No. A1317609
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2856-63-5 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2856-63-5
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001898
InChI Key MRDUURPIPLIGQX-UHFFFAOYSA-N
Synonym 2-chlorobenzyl cyanide, 2-chlorophenylacetonitrile, 2-2-chlorophenyl acetonitrile, o-chlorobenzyl cyanide, benzeneacetonitrile, 2-chloro, 2-chlorobenzeneacetonitrile, 2-chlorobenzylcyanide, 2-chlorophenyl acetonitrile, o-chlorophenyl acetonitrile, acetonitrile, o-chlorophenyl
PubChem CID 76112
IUPAC Name 2-(2-chlorophenyl)acetonitrile
SMILES C1=CC=C(C(=C1)CC#N)Cl

Specifications

Melting Point 21°C to 24°C
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 10 g
UN Number UN3439
Beilstein 2043683
Formula Weight 151.6
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenylacetonitrile
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H300-H311-H331-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 220-669-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.