Learn More
Medchemexpress LLC 2-chloropropane-1,3-diyl distearate-d5 | 1329796-49-7 | 99.1% | 648.49 g/mol | C39H70D5ClO4 | 5 MG
SDP
Catalog No. 5000284104
Encompass_Preferred
Deuterium-labeled stable isotope standard intended as a tracer and internal standard for quantitative analytical methods. Supplied with supporting documentation to aid analytical validation and handling; intended for research use only. Molecular formula C39H70D5ClO4, molecular weight 648.49 g/mol, CAS 1329796-49-7.
  • Useful as tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium labeling enables clear mass separation from native analytes.
  • High purity supports accurate and reproducible quantitation.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Available in mg-scale quantities suitable for analytical workflows.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-84104 Supplier Medchemexpress LLC Supplier No. HYW585809S5MG
Deuterium-labeled stable isotope standard intended as a tracer and internal standard for quantitative analytical methods. Supplied with supporting documentation to aid analytical validation and handling; intended for research use only. Molecular formula C39H70D5ClO4, molecular weight 648.49 g/mol, CAS 1329796-49-7.
  • Useful as tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium labeling enables clear mass separation from native analytes.
  • High purity supports accurate and reproducible quantitation.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Available in mg-scale quantities suitable for analytical workflows.
  • Intended for research use only; not for human or clinical use.