Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloropropane-1,3-diyl distearate-d5 | 1329796-49-7 | MFCD30718237 | 99.1% | 648.49 | C39H70D5ClO4 | 1 MG
SDP

Catalog No. 5000284687
Encompass_Preferred

2-Chloropropane-1,3-diyl distearate-d5 is a deuterium-labeled lipid derivative supplied in small quantities for analytical research. It is designed for use as a tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS, and is provided with supporting documentation for analytical workflows.

  • Deuterium-labeled tracer and internal standard.
  • High purity (99.1%).
  • Molecular formula C39H70D5ClO4 and molecular weight 648.49.
  • CAS number 1329796-49-7 for unambiguous identification.
  • Available in small-mass sizes suitable for analytical use (1 mg-100 mg).
  • Supplied with data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-84687 Supplier Medchemexpress LLC Supplier No. HYW585809S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloropropane-1,3-diyl distearate-d5 is a deuterium-labeled lipid derivative supplied in small quantities for analytical research. It is designed for use as a tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS, and is provided with supporting documentation for analytical workflows.

  • Deuterium-labeled tracer and internal standard.
  • High purity (99.1%).
  • Molecular formula C39H70D5ClO4 and molecular weight 648.49.
  • CAS number 1329796-49-7 for unambiguous identification.
  • Available in small-mass sizes suitable for analytical use (1 mg-100 mg).
  • Supplied with data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.