Learn More
Medchemexpress LLC 2-chloropropane-1,3-diyl distearate-d5 (major) | 1329796-49-7 | 99.1% | 648.49 g/mol | C39H70D5ClO4 | 1 MG
SDP
Catalog No. 5000284223
Encompass_Preferred
2-Chloropropane-1,3-diyl distearate-d5 (major) is a deuterium-labeled analogue of 2-chloropropane-1,3-diyl distearate supplied as a high-purity stable-isotope standard for laboratory research. It is intended for use as a tracer or internal standard in analytical methods such as LC-MS, GC-MS, and NMR, and is provided in milligram quantities for quantitative and qualitative studies. Research use only.
  • Deuterium-labeled tracer for isotope dilution assays.
  • High purity suitable for analytical reference standards.
  • Verified molecular formula and mass for accurate identification.
  • Supplied in small milligram quantities for method development.
  • Provided with batch-specific certificate of analysis and documentation.
Catalog No. 50-002-84223 Supplier Medchemexpress LLC Supplier No. HYW585809S11MG
2-Chloropropane-1,3-diyl distearate-d5 (major) is a deuterium-labeled analogue of 2-chloropropane-1,3-diyl distearate supplied as a high-purity stable-isotope standard for laboratory research. It is intended for use as a tracer or internal standard in analytical methods such as LC-MS, GC-MS, and NMR, and is provided in milligram quantities for quantitative and qualitative studies. Research use only.
  • Deuterium-labeled tracer for isotope dilution assays.
  • High purity suitable for analytical reference standards.
  • Verified molecular formula and mass for accurate identification.
  • Supplied in small milligram quantities for method development.
  • Provided with batch-specific certificate of analysis and documentation.