Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropropionamide, 99%, Thermo Scientific™

Catalog No. AC185210100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC185210100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC185210100 Supplier Thermo Scientific Chemicals Supplier No. 185210100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27816-36-0
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.54
MDL Number MFCD00008016
InChI Key OEZPDHRXGCLGKB-UHFFFAOYNA-N
Synonym 2-chloropropionamide, propanamide, 2-chloro, a-chloropropionamide, 2-chloro-propanamide, #, propanamide,2-chloro, acmc-1ckh3, 2-chloropropionic acid amide, oezpdhrxgclgkb-uhfffaoysa
PubChem CID 94385
IUPAC Name 2-chloropropanamide
SMILES CC(Cl)C(N)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 76°C to 80°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula CH3CH(Cl)CONH2
Quantity 10 g
Beilstein 02, 249
Formula Weight 107.54
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloropropionamide, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.