Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-2-Chloropropionic Acid

Catalog No. 3068001G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-680-01G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 30-680-01G Supplier Sigma Aldrich Supplier No. 3068001G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-2-Chloropropionic acid can be used as a starting material in the synthesis of: Thiolactic acid, a key intermediate, which is used for the preparation of 2(R),5(S)-oxathiolanones by reacting with pivalaldehyde (R)-2-chloropropionyl chloride, a key intermediate, which is utilized in the synthesis of (R)-4,5-dihydro-5-methylpyridazin-3(2H)-one derivative with pyrazolopyridine ring as a potential phosphodiesterase inhibitor 2-Chloropropionamide derivatives as protein disulfide isomerase (PID) inhibitors.

Specifications

CAS 27126-76-7
Melting Point 131°C to 137°C
Molecular Formula C13H13IO4S
Refractive Index n20/D 1.4345
Linear Formula CH3C6H4SO3I(OH)C6H5
Quantity 25 g
MDL Number MFCD00011547
Synonym HTIB; Hydroxy(4-methylbenzenesulfonato-O)phenyliodine; Hydroxy(phenyl)iodo tosylate; Iodosobenzene-I-mono-p-toluenesulfonate; Koser's reagent; NSC 294176; Phenyliodosyl hydroxide tosylate; [Hydroxy(4-toluenesulfonato)iodo]benzene
Molecular Weight (g/mol) 392.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.