Learn More
Medchemexpress LLC 2-chloropyrazine | 14508-49-7 | MFCD00006124 | 99.7% | 114.53 | C4H3ClN2 | 50g
SDP
Catalog No. 5000268179
Encompass_Preferred
Chloropyrazine (Pyrazin-2-yl chloride) is a pyrazine-based compound that can be used for the synthesis of active compounds Additionally Chloropyrazine can be used as an electrolyte additive in the research of lithium-sulfur batteries[1][2]
Catalog No. 50-002-68179 Supplier Medchemexpress LLC Supplier No. HYY010950G
Chloropyrazine (Pyrazin-2-yl chloride) is a pyrazine-based compound that can be used for the synthesis of active compounds Additionally Chloropyrazine can be used as an electrolyte additive in the research of lithium-sulfur batteries[1][2]