Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193731 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25601G 1 g
C2560-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25601G Supplier TCI America Supplier No. C25601G
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2-Chloropyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 381248-04-0
Melting Point 160°C
Color White-Yellow
Molecular Formula C5H5BClNO2
Quantity 1 g
MDL Number MFCD03094997
Synonym 2-chloropyridine-3-boronic acid, 2-chloropyridin-3-yl boronic acid, 2-chloro-3-pyridylboronic acid, 2-chloro-3-pyridineboronic acid, 2-chloropyridine-3-boronicacid, 2-chloropyridin-3-yl-3-boronic acid, 2-chloro-3-pyridyl boronic acid, boronic acid, 2-chloro-3-pyridinyl, pubchem5078
InChI Key VRDAOVQZVXYRNH-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CN=C1Cl
IUPAC Name (2-chloropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 157.36
PubChem CID 2762704
Formula Weight 157.36
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.