Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C22885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2288-5G 5 g
C22881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2288-5G Supplier TCI America Supplier No. C22885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 444120-91-6 | C5H5BClNO2 | 157.36 g/mol

Specifications

CAS 444120-91-6
Melting Point 167°C
Color White-Yellow
Molecular Formula C5H5BClNO2
MDL Number MFCD03094998
Quantity 5 g
Synonym 2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid
InChI Key WPAPNCXMYWRTTL-UHFFFAOYSA-N
SMILES B(C1=CN=C(C=C1)Cl)(O)O
IUPAC Name (6-chloropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 157.36
PubChem CID 3845034
Formula Weight 157.36
Percent Purity 96.0 to 113.0%
Grade Research
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.