Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropyridine, 99%, MP Biomedicals™

Catalog No. ICN15065325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15065325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15065325 Supplier MP Biomedicals, Inc Supplier No. 0215065325

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109-09-1
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.544
InChI Key OKDGRDCXVWSXDC-UHFFFAOYSA-N
Synonym o-chloropyridine, pyridine, 2-chloro, alpha-chloropyridine, .alpha.-chloropyridine, unii-8hmd45ayej, 2-chloro-pyridine, ccris 1724, 8hmd45ayej, 6-chloro-pyridine, dsstox_cid_4810
PubChem CID 7977
ChEBI CHEBI:39174
IUPAC Name 2-chloropyridine
SMILES C1=CC=NC(=C1)Cl

Specifications

Assay Percent Range 99%
Quantity 25 g
Formula Weight 113.5
Physical Form Liquid
Chemical Name or Material 2-Chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.