Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloropyrimidine-5-carboxylic acid | 374068-01-6 | 99.7% | C5H3ClN2O2 | 1 G
SDP

Catalog No. 5000434290
Encompass_Preferred

2-Chloropyrimidine-5-carboxylic acid is a drug intermediate used for the synthesis of various active compounds. This product is for research use only and not intended for patient use.

  • Solid appearance
  • White to light yellow color
  • Molecular weight of 158.54
  • Purity of 99.7%
  • Store at 4°C under nitrogen

Catalog No. 50-004-34290 Supplier Medchemexpress LLC Supplier No. HY691421G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloropyrimidine-5-carboxylic acid is a drug intermediate used for the synthesis of various active compounds. This product is for research use only and not intended for patient use.

  • Solid appearance
  • White to light yellow color
  • Molecular weight of 158.54
  • Purity of 99.7%
  • Store at 4°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.