Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloropyrimidine-5-carboxylic acid | 374068-01-6 | 99.7% | C5H3ClN2O2 | 5 G
SDP

Catalog No. 5000434698
Encompass_Preferred

2-Chloropyrimidine-5-carboxylic acid is a drug intermediate used for the synthesis of various active compounds. It is supplied as a solid, typically white to light yellow in color.

  • Molecular weight: 158.54
  • Chemical formula: C5H3ClN2O2
  • CAS number: 374068-01-6
  • Appearance: White to light yellow solid
  • Recommended storage at 4°C under nitrogen
  • Long-term storage in solvent: -80°C for 6 months or -20°C for 1 month under nitrogen
  • High solubility in DMSO (100 mg/mL)
  • Can be dissolved in various in vivo solutions at ≥ 5 mg/mL

Catalog No. 50-004-34698 Supplier Medchemexpress LLC Supplier No. HY691425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloropyrimidine-5-carboxylic acid is a drug intermediate used for the synthesis of various active compounds. It is supplied as a solid, typically white to light yellow in color.

  • Molecular weight: 158.54
  • Chemical formula: C5H3ClN2O2
  • CAS number: 374068-01-6
  • Appearance: White to light yellow solid
  • Recommended storage at 4°C under nitrogen
  • Long-term storage in solvent: -80°C for 6 months or -20°C for 1 month under nitrogen
  • High solubility in DMSO (100 mg/mL)
  • Can be dissolved in various in vivo solutions at ≥ 5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.