Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloropyrimidine-5-carboxylic acid | 374068-01-6 | 99.96% | C5H3ClN2O2 | 25 G
SDP

Catalog No. 5000358457
Encompass_Preferred

2-Chloropyrimidine-5-carboxylic acid is a versatile drug intermediate utilized in the synthesis of various active compounds. This product is provided as a solid, typically white to light yellow in appearance, and requires specific storage conditions to maintain its integrity.

  • Solid, white to light yellow appearance
  • Store at 4°C under nitrogen
  • Soluble in DMSO at 100 mg/mL (requires sonication)
  • For stock solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-003-58457 Supplier Medchemexpress LLC Supplier No. 25GHY69142
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Chloropyrimidine-5-carboxylic acid is a versatile drug intermediate utilized in the synthesis of various active compounds. This product is provided as a solid, typically white to light yellow in appearance, and requires specific storage conditions to maintain its integrity.

  • Solid, white to light yellow appearance
  • Store at 4°C under nitrogen
  • Soluble in DMSO at 100 mg/mL (requires sonication)
  • For stock solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.