Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloroquinoxaline, 97%

Catalog No. AC136190050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC136190050 5 g Glass bottle
AC136190010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC136190050 Supplier Thermo Scientific Chemicals Supplier No. 136190050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1448-87-9 | C8H5ClN2 | 164.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448-87-9
Molecular Formula C8H5ClN2
Molecular Weight (g/mol) 164.59
MDL Number MFCD00043907
InChI Key BYHVGQHIAFURIL-UHFFFAOYSA-N
Synonym quinoxaline, 2-chloro, unii-e9x60z79bg, chloroquinoxaline, quinoxalyl chloride, 2-chloro quinoxaline, 2-chloro-quinoxaline, 2-chloranylquinoxaline, 2-chloro-benzopyrazine, pubchem15447, 2-chloroquinoxaline
PubChem CID 238938
IUPAC Name 2-chloroquinoxaline
SMILES ClC1=CN=C2C=CC=CC2=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 48.0°C to 50.0°C
Color White to Gray
Boiling Point 100.0°C (1.4 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 164.6
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloroquinoxaline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 215-905-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.