Learn More
Chem-Impex International, Inc. 2-Chlorotrityl chloride resin (1.0 - 1.6 mmol/g, 100 - 200 mesh) | 42074-68-0 | MFCD00161760 | 25G
Catalog No. 50497933
Encompass_Preferred
2-Chlorotrityl chloride resin (1.0 - 1.6 mmol/g, 100 - 200 mesh), 42074-68-0, MFCD00161760, 25G
Catalog No. 50-497-933 Supplier Chem-Impex International, Inc. Supplier No. 0349825G
2-Chlorotrityl chloride resin (1.0 - 1.6 mmol/g, 100 - 200 mesh), 42074-68-0, MFCD00161760, 25G