Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chlorotrityl chloride | 42074-68-0 | 98.0% | 313.23 | 1 G
SDP

Catalog No. 5000366567
Encompass_Preferred

2-Chlorotrityl chloride, also known as (Chloro(2-chlorophenyl)methylene)dibenzene, is a biochemical reagent. It is suitable for use as a biological material or organic compound in life science related research. This product is intended for research use only and is not for medical or patient use.

  • Purity of 98.0%
  • Molecular weight of 313.23
  • Appearance as an off-white to light yellow solid
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 25 mg/mL
  • Soluble in vivo at ≥ 2.5 mg/mL in 10% DMSO and 90% corn oil

Catalog No. 50-003-66567 Supplier Medchemexpress LLC Supplier No. HYW0113311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chlorotrityl chloride, also known as (Chloro(2-chlorophenyl)methylene)dibenzene, is a biochemical reagent. It is suitable for use as a biological material or organic compound in life science related research. This product is intended for research use only and is not for medical or patient use.

  • Purity of 98.0%
  • Molecular weight of 313.23
  • Appearance as an off-white to light yellow solid
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 25 mg/mL
  • Soluble in vivo at ≥ 2.5 mg/mL in 10% DMSO and 90% corn oil

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.