Learn More
Bachem 2-Chlorotrityl chloride resin (200-400 mesh, 1.5-1.9 mmol/g)
SDP
Catalog No. 50258901
Encompass_Preferred
2-Chlorotrityl chloride resin (200-400 mesh, 1.5-1.9 mmol/g), 934816-82-11, n/a, Mr n/a, 5g. 2-ClTrt resin, 2-CTC resin. A very acid sensitive resin suitable for the synthesis of protected peptide fragments by the Fmoc-strategy. ClTrt resin is in particular suitable for the preparation of C-terminal prolyl and cysteinyl peptides (please see 2-Chlorotrityl Chloride Resin-Linked Amino Acids). Release from this resin is achieved by using 1-50 % TFA in CH2Cl2 containing 8 % triisopropylsilane, AcOH/CF3CH2OH/CH2Cl2, 0.5 % TFA or hexafluoroisopropanol/CH2Cl2. ClTrt resin has been used in cyclization reactions under Heck reaction conditions, the solid phase synthesis of b-peptides via the Arndt-Eistert homologation of Fmoc-protected amino acid diazoketones and in Mannich reactions of alkynes, secondary amines and aldehydes
Catalog No. 50-258-901 Supplier Bachem Supplier No. 4025425.0005
2-Chlorotrityl chloride resin (200-400 mesh, 1.5-1.9 mmol/g), 934816-82-11, n/a, Mr n/a, 5g. 2-ClTrt resin, 2-CTC resin. A very acid sensitive resin suitable for the synthesis of protected peptide fragments by the Fmoc-strategy. ClTrt resin is in particular suitable for the preparation of C-terminal prolyl and cysteinyl peptides (please see 2-Chlorotrityl Chloride Resin-Linked Amino Acids). Release from this resin is achieved by using 1-50 % TFA in CH2Cl2 containing 8 % triisopropylsilane, AcOH/CF3CH2OH/CH2Cl2, 0.5 % TFA or hexafluoroisopropanol/CH2Cl2. ClTrt resin has been used in cyclization reactions under Heck reaction conditions, the solid phase synthesis of b-peptides via the Arndt-Eistert homologation of Fmoc-protected amino acid diazoketones and in Mannich reactions of alkynes, secondary amines and aldehydes