Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Chlorotrityl chloride resin (100-200 mesh), 1% DVB, 5G, 934816-82-7, MFCD00161760
Catalog No. NC1255133
Encompass
An extremely versatile, acid-labile resin for the solid phase immobilization of carboxylic acids, alcohols, phenols and amines, imidazoles and hydroxylamines. Release of these functionalities is generally achieved using 1-50% TFA in DCM containing 5% TIS, although carboxylic acids can also be cleaved from this support with AcOH/TFE/DCM, 0.5% TFA in DCM, or HFIP in DCM.
Catalog No. NC1255133 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8550170005
An extremely versatile, acid-labile resin for the solid phase immobilization of carboxylic acids, alcohols, phenols and amines, imidazoles and hydroxylamines. Release of these functionalities is generally achieved using 1-50% TFA in DCM containing 5% TIS, although carboxylic acids can also be cleaved from this support with AcOH/TFE/DCM, 0.5% TFA in DCM, or HFIP in DCM.