Learn More
Chem-Impex International, Inc. 2-CHLOROTRITYL CHLORIDE RES25G
Catalog No. NC2221688
Encompass_Preferred
2-Chlorotrityl chloride resin (0.4 - 0.8 mmol/g, 100 - 200 mesh) 25g CAS#42074-68-0 MFCD00161760
This item is not returnable. View return policy
Catalog No. NC2221688 Supplier Chem-Impex International, Inc. Supplier No. 3331025G
2-Chlorotrityl chloride resin (0.4 - 0.8 mmol/g, 100 - 200 mesh) 25g CAS#42074-68-0 MFCD00161760