Learn More
Cole-Parmer Partlow 711000000021 2 Chnl Chart Recorder Std Cover 90-115 Vac
Catalog No. 50628403 Shop All Cole-Parmer Products
Encompass
Proven recording reliability that became the industry standard! Capable of measuring, displaying, recording and controlling up to two process variables from a variety of inputs. All user entered data can be protected against unauthorized changes. Straightforward programming allows you to configure your recorder with a logical step-by-step process using a simple keypad.
Catalog No. 50-628-403 Supplier Cole-Parmer Supplier No. 4900193
Proven recording reliability that became the industry standard! Capable of measuring, displaying, recording and controlling up to two process variables from a variety of inputs. All user entered data can be protected against unauthorized changes. Straightforward programming allows you to configure your recorder with a logical step-by-step process using a simple keypad.