Learn More
Cayman Chemical 2-cIs-HexadecenoIc AcId 10mg
SDP
Catalog No. 502951554
Encompass_Preferred
An unusual fatty acid that acylates some Myxococcus species, a group of gram-negative bacteria, found mainly in soil and also common to marine and freshwater systems with quorum sensing capabilities
Catalog No. 50-295-1554 Supplier Cayman Chemical Supplier No. 1113310 MG
An unusual fatty acid that acylates some Myxococcus species, a group of gram-negative bacteria, found mainly in soil and also common to marine and freshwater systems with quorum sensing capabilities