Learn More
Eurofins Discoverx 2-Cl-IB-MECA, 10 mg. (92-1045)
Supplier:  Eurofins Discoverx 921045
Encompass
2-Cl-IB-MECA is a potent and selective agonist of A3 adenosine receptor (ADORA3, Ki=0.33 nM). Selective over A1 and A2 receptors by 2500- and 1400-fold, respectively. ADORA3 has been implicated in cardioprotective functions during ischemia, neuroprotection and neurodegeneration, as well as cell proliferation and cell death mediation. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9589