Learn More
Waters Corp Waters Two-Column Regeneration Valve Kit, 2 Position, Ten-port, Automated Switching
Catalog No. 50791629 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 2 position regeneration automated switching valve is designed for use with Alliance HPLC series Column Heater and Column Heater/Cooler modules. It provides a ten-port, two-position configuration to automate off-line alternate column regeneration.
  • Automates off-line alternate column regeneration to significantly increase overall sample throughput.
  • Includes a ten-port, two-position valve kit.
  • Pre-installed on a mounting plate for quick and easy installation into Column Heater or Column Heater/Cooler Modules.
  • Comes with tubing and fittings for connecting to column(s) and other external devices.
  • Connects to an external column regeneration pump.
  • Programmable from the instrument’s front panel or remotely via Empower™ or MassLynx™ software.
Catalog No. 50-791-629 Supplier Waters Corp Supplier No. 205000163
This 2 position regeneration automated switching valve is designed for use with Alliance HPLC series Column Heater and Column Heater/Cooler modules. It provides a ten-port, two-position configuration to automate off-line alternate column regeneration.
  • Automates off-line alternate column regeneration to significantly increase overall sample throughput.
  • Includes a ten-port, two-position valve kit.
  • Pre-installed on a mounting plate for quick and easy installation into Column Heater or Column Heater/Cooler Modules.
  • Comes with tubing and fittings for connecting to column(s) and other external devices.
  • Connects to an external column regeneration pump.
  • Programmable from the instrument’s front panel or remotely via Empower™ or MassLynx™ software.