Learn More
Cayman Chemical 2-Coumarnon-1-L 5mg
SDP
Catalog No. 502937715
Encompass_Preferred
A chemiluminescent probe; em = 442 nm in the presence of a base and oxygen
Catalog No. 50-293-7715 Supplier Cayman Chemical Supplier No. 273895 MG
A chemiluminescent probe; em = 442 nm in the presence of a base and oxygen