Learn More
Medchemexpress LLC 2-CTC Resin | 934816-82-7 | 100 G
SDP
Catalog No. 5000352958
Encompass_Preferred
2-CTC Resin is a solid-phase synthesis carrier recognized for its versatility in large-scale peptide production. It prevents racemization during initial amino acid incorporation and minimizes diketopiperazine formation. The resin has been used to investigate metabolic disorders in prostate cancer cells and can form cyclic peptides from terminal residues and adenine nucleotides.
  • Solid-phase synthesis carrier
  • Prevents racemization during amino acid incorporation
  • Minimizes diketopiperazine formation
  • Versatile for large-scale peptide production
  • Forms covalent ester bonds with amino acids for efficient immobilization
Catalog No. 50-003-52958 Supplier Medchemexpress LLC Supplier No. HY142104100G
2-CTC Resin is a solid-phase synthesis carrier recognized for its versatility in large-scale peptide production. It prevents racemization during initial amino acid incorporation and minimizes diketopiperazine formation. The resin has been used to investigate metabolic disorders in prostate cancer cells and can form cyclic peptides from terminal residues and adenine nucleotides.
  • Solid-phase synthesis carrier
  • Prevents racemization during amino acid incorporation
  • Minimizes diketopiperazine formation
  • Versatile for large-scale peptide production
  • Forms covalent ester bonds with amino acids for efficient immobilization