Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-2,2-dimethylacetic acid, 95%, Thermo Scientific™

Catalog No. AAH6673403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6673403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6673403 Supplier Thermo Scientific Chemicals Supplier No. H6673403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22426-30-8
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD11100210
InChI Key DZDVNAVPCCNJRK-UHFFFAOYSA-N
Synonym 2-cyano-2,2-dimethylacetic acid, 2-cyano-2-methyl-propanoic acid, cyanomethylpropanoicacid, 2-cyanoisobutyric acid, dimethylcyanoacetic acid, propanoic acid, 2-cyano-2-methyl
PubChem CID 11367091
IUPAC Name 2-cyano-2-methylpropanoic acid
SMILES CC(C)(C#N)C(O)=O

Specifications

Melting Point 56°C to 61°C
Color White
Boiling Point 132°C to 135°C (12 mmHg)
Quantity 1 g
UN Number UN2923
Sensitivity Moisture sensitive
Formula Weight 113.11
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 2-Cyano-2,2-dimethylacetic acid
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.