Learn More
Medchemexpress LLC 2-cyano-3-[5-(2,5-dichlorophenyl)-2-furanyl]-N-5-quinolinyl-2-propenamide | 304896-28-4 | 99.6% | 434.27 | C23H13Cl2N3O2 | 100 MG
SDP
Catalog No. 5000211935
Encompass_Preferred
AGK2 is a selective small-molecule inhibitor of SIRT2 used to probe sirtuin biology and apoptosis-related pathways in biochemical and cellular studies. It is cell permeable and reported to inhibit SIRT2 with an IC50 of 3.5 μM, while showing weaker activity against SIRT1 and SIRT3.
Catalog No. 50-002-11935 Supplier Medchemexpress LLC Supplier No. HY100578100MG
AGK2 is a selective small-molecule inhibitor of SIRT2 used to probe sirtuin biology and apoptosis-related pathways in biochemical and cellular studies. It is cell permeable and reported to inhibit SIRT2 with an IC50 of 3.5 μM, while showing weaker activity against SIRT1 and SIRT3.