Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-3-methylpyridine, 98%

Catalog No. p-7047568
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1945606 5 g
AAL1945614 25 g
AAL1945622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1945606 Supplier Thermo Scientific Chemicals Supplier No. L1945606
Only null left
Add to Cart

CAS: 20970-75-6 | C7H6N2 | 118.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20970-75-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00190585
InChI Key WBXZCDIZXWDPBL-UHFFFAOYSA-N
Synonym 2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
PubChem CID 819928
IUPAC Name 3-methylpyridine-2-carbonitrile
SMILES CC1=C(N=CC=C1)C#N

Specifications

Melting Point 84°C to 87°C
Boiling Point 141°C (38 mmHg)
Flash Point 115°C (239°F)
Odor Characteristic
Quantity 5 g
Beilstein 112001
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 2-Cyano-3-methylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-131-3
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.