Learn More
eMolecules​ 5331-42-0 | 2-CYANO-3-PHENYLPROPIONIC ACID | AstaTech | MFCD00205073 | 175.187 | C10H9NO2 | 97.000 | OC(=O)C(Cc1ccccc1)C#N | 5g | 222803423
Catalog No. 501573760 Shop All eMolecules Inc. Products
Encompass_Preferred
2-CYANO-3-PHENYLPROPIONIC ACID | AstaTech | 5331-42-0 | MFCD00205073 | 175.187 | C10H9NO2 | 97.000 | OC(=O)C(Cc1ccccc1)C#N | 5g | 222803423
Catalog No. 50-157-3760 Supplier eMolecules​ Supplier No. 222803423
2-CYANO-3-PHENYLPROPIONIC ACID | AstaTech | 5331-42-0 | MFCD00205073 | 175.187 | C10H9NO2 | 97.000 | OC(=O)C(Cc1ccccc1)C#N | 5g | 222803423