Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%

Catalog No. AAH61451MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61451MD 250 mg
AAH6145103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61451MD Supplier Thermo Scientific Chemicals Supplier No. H61451MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1218790-84-1 | C8H5BF3NO2 | 214.938 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218790-84-1
Molecular Formula C8H5BF3NO2
Molecular Weight (g/mol) 214.938
InChI Key DELAZGQTKSRLPA-UHFFFAOYSA-N
Synonym 2-cyano-4-trifluoromethyl phenylboronic acid, 2-cyano-4-trifluoromethyl phenyl boronic acid, acmc-209ail, 2-cyano-4-trifluoromethyl phenyl boronicacid
PubChem CID 53216599
IUPAC Name [2-cyano-4-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(F)(F)F)C#N)(O)O

Specifications

Quantity 250 mg
Formula Weight 214.94
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Cyano-4-(trifluoromethyl)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.