Learn More
Medchemexpress LLC 2-cyano-N-{[(3,4-dichlorophenyl)carbamothioyl]amino}acetamide | 656222-54-7 | MFCD00119122 | 99.8% | 303.17 | C10H8Cl2N4OS | 25MG
SDP
Catalog No. 5000237141
Encompass_Preferred
iKIX1 is a small-molecule antifungal research compound that disrupts the interaction between the KIX domain of the mediator subunit CgGal11A and the activation domain of CgPdr1, and it resensitizes azole-resistant Candida glabrata in vitro. The compound has formula C10H8Cl2N4OS and a molecular weight of 303.17 g·mol⁻¹; it is supplied for research use only.
  • Inhibits KIX-activation domain interaction (IC50 190.2 μM; Ki 18 μM).
  • Resensitizes azole-resistant Candida glabrata in vitro.
  • Supplied for research use only; not for clinical use.
  • High purity (99.8% as reported by manufacturer).
  • Stable as powder at -20°C for up to 3 years; in solvent stored at -80°C (6 months) or -20°C (1 month).
  • Available as solid and as DMSO solution formulations for convenient handling.
Catalog No. 50-002-37141 Supplier Medchemexpress LLC Supplier No. HY12495225MG
iKIX1 is a small-molecule antifungal research compound that disrupts the interaction between the KIX domain of the mediator subunit CgGal11A and the activation domain of CgPdr1, and it resensitizes azole-resistant Candida glabrata in vitro. The compound has formula C10H8Cl2N4OS and a molecular weight of 303.17 g·mol⁻¹; it is supplied for research use only.
  • Inhibits KIX-activation domain interaction (IC50 190.2 μM; Ki 18 μM).
  • Resensitizes azole-resistant Candida glabrata in vitro.
  • Supplied for research use only; not for clinical use.
  • High purity (99.8% as reported by manufacturer).
  • Stable as powder at -20°C for up to 3 years; in solvent stored at -80°C (6 months) or -20°C (1 month).
  • Available as solid and as DMSO solution formulations for convenient handling.