Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-cyano-N-{[(3,4-dichlorophenyl)carbamothioyl]amino}acetamide | 656222-54-7 | MFCD00119122 | 99.8% | 303.17 g/mol | C10H8Cl2N4OS | 100MG
SDP

Catalog No. 5000235492
Encompass_Preferred

iKIX1 is an antifungal research compound that resensitizes drug-resistant Candida glabrata to azole antifungals by inhibiting the interaction between the KIX domain of mediator subunit CgGal11A and the activation domain of CgPdr1. It is used for in vitro studies of multidrug resistance and fungal infection models.

  • Molecular formula: C10H8Cl2N4OS.
  • Molecular weight: 303.17 g/mol.
  • Purity: 99.8% (HPLC).
  • Appearance: off-white to light yellow solid.
  • Solubility: DMSO 83.33 mg/mL (≈274.9 mM); may require ultrasonic.
  • Storage: powder -20°C (up to 3 years); in solution -80°C (6 months), -20°C (1 month).
  • Available sizes: solid quantities from milligram to gram scale and 10 mM solutions in DMSO.

Catalog No. 50-002-35492 Supplier Medchemexpress LLC Supplier No. HY124952100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

iKIX1 is an antifungal research compound that resensitizes drug-resistant Candida glabrata to azole antifungals by inhibiting the interaction between the KIX domain of mediator subunit CgGal11A and the activation domain of CgPdr1. It is used for in vitro studies of multidrug resistance and fungal infection models.

  • Molecular formula: C10H8Cl2N4OS.
  • Molecular weight: 303.17 g/mol.
  • Purity: 99.8% (HPLC).
  • Appearance: off-white to light yellow solid.
  • Solubility: DMSO 83.33 mg/mL (≈274.9 mM); may require ultrasonic.
  • Storage: powder -20°C (up to 3 years); in solution -80°C (6 months), -20°C (1 month).
  • Available sizes: solid quantities from milligram to gram scale and 10 mM solutions in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.