Learn More
Medchemexpress LLC 2-cyano-N-{[(3,4-dichlorophenyl)carbamothioyl]amino}acetamide | 656222-54-7 | MFCD00119122 | 99.8% | 303.17 g·mol⁻¹ | C10H8Cl2N4OS | 50MG
SDP
Catalog No. 5000235569
Encompass_Preferred
iKIX1 is a small-molecule antifungal research compound that resensitizes drug-resistant Candida glabrata to azole antifungals in vitro by inhibiting the interaction between the KIX domain of mediator subunit CgGal11A and the activation domain of CgPdr1. It is provided in multiple solid pack sizes and as a 10 mM solution in DMSO for in vitro studies; reported purity is 99.76%.
  • Resensitizes drug-resistant Candida glabrata to azole antifungals in vitro.
  • Inhibits the CgGal11A-CgPdr1 protein-protein interaction.
  • Available as solid (5-500 mg) and as a 10 mM solution in DMSO.
  • High reported purity (99.76%) suitable for biochemical assays.
  • Molecular formula C10H8Cl2N4OS; molecular weight 303.17 g·mol⁻¹.
Catalog No. 50-002-35569 Supplier Medchemexpress LLC Supplier No. HY12495250MG
iKIX1 is a small-molecule antifungal research compound that resensitizes drug-resistant Candida glabrata to azole antifungals in vitro by inhibiting the interaction between the KIX domain of mediator subunit CgGal11A and the activation domain of CgPdr1. It is provided in multiple solid pack sizes and as a 10 mM solution in DMSO for in vitro studies; reported purity is 99.76%.
  • Resensitizes drug-resistant Candida glabrata to azole antifungals in vitro.
  • Inhibits the CgGal11A-CgPdr1 protein-protein interaction.
  • Available as solid (5-500 mg) and as a 10 mM solution in DMSO.
  • High reported purity (99.76%) suitable for biochemical assays.
  • Molecular formula C10H8Cl2N4OS; molecular weight 303.17 g·mol⁻¹.