Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide 98.0+%, TCI America™
SDP

Catalog No. C32175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32175G 5 g
C32171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32175G Supplier TCI America Supplier No. C32175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24522-30-3
Molecular Formula C10H7F3N2O
Molecular Weight (g/mol) 228.174
MDL Number MFCD05669245
InChI Key JBNCFFDGYDZEEN-UHFFFAOYSA-N
PubChem CID 3823803
IUPAC Name 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
SMILES C1=CC(=CC=C1C(F)(F)F)NC(=O)CC#N

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 5 g
Formula Weight 228.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.