Learn More
Selleck Chemical LLC 2-cyano-Pyrimidine
SDP
Catalog No. 5000003024
Encompass_Preferred
2-cyano-Pyrimidine is a cathepsin K inhibitor with an IC50 of 170 nM
Catalog No. 50-000-03024 Supplier Selleck Chemical LLC Supplier No. S561125MG
2-cyano-Pyrimidine is a cathepsin K inhibitor with an IC50 of 170 nM