Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. c26565g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26565G 5 g
C265625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26565G Supplier TCI America Supplier No. C26565G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 612-13-5
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00019745
InChI Key ZSHNOXOGXHXLAV-UHFFFAOYSA-N
Synonym 2-(Chloromethyl)benzonitrile, alpha-Chloro-o-tolunitrile
PubChem CID 69152
IUPAC Name 2-(chloromethyl)benzonitrile
SMILES ClCC1=CC=CC=C1C#N

Specifications

Melting Point 61°C
Color White
Boiling Point 252°C
Quantity 5 g
UN Number 2923
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanobenzyl Chloride
EINECSNumber (3)-1810
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.