Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Cyanobenzylzinc Bromide Solution

Catalog No. 52032250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-032-250ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 52-032-250ML Supplier Sigma Aldrich Supplier No. 52032250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Cyanobenzylzinc bromide can be used as a reactant: In the metal-catalyzed Negishi cross-coupling reactions to prepare aryl or heteroaryl derivatives via carbon-carbon bond formation. To synthesize 4-(2-Cyanobenzyl)-3'sa-(trifluoromethyl)biphenyl by reacting with aryl nonaflate in the presence of Pd(dba)2 as a catalyst. To prepare 2-[(2-cyanophenyl)methyl] benzamide by treating with 2-iodobenzamide using a nickel catalyst.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 15186-48-8
Density 1.045 g/mL at 25°C
Boiling Point 139°C
Molecular Formula C6H10O3
Linear Formula C6H10O3
MDL Number MFCD00269682
Quantity 500 mg
Molecular Weight (g/mol) 130.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.