Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Cyanoethyl N,N,N',N'-tetraisopropylphosphorodiamidite

Catalog No. 3059951G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-599-51G 1 g
30-599-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-599-51G Supplier Sigma Aldrich Supplier No. 3059951G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-cyanoethyl n,n,nα,nα-tetraisopropylphosphordiamidite was used:
  • In preparation of prosphoramidite reagent, required for synthesis of 12-mer oligodeoxynucleotide
  • As phosphorylating agent in synthesis of 1,2-diacyl-sn-glycerophosphatidylserine
  • In situ preparation of deoxyribonucleoside phosphoramidites
  • In preparation of 2α-deoxy-2α-fluoro-3α-o-(α-cyanoethyl-n,n-diisopropylphosphoramidic)-5α-o-(4-methoxytrityl)-4α-thio-α-d-arabinouridine and 1-(3-o-(α-cyanoethyl-n,n-diisopropylphosphoramidic)-2-deoxy-2-fluoro-5-o-(4,4α-dimethoxytrityl)-4-thio-α-d-arabinofuranosyl)-thymine
  • As reagent for synthesizing phosphitylated nucleotides

Specifications

CAS 102691-36-1
Density 0.949 g/mL (at 25°C (literature))
Boiling Point 100°C (0.5 mmHg)
Molecular Formula C15H32N3OP
Refractive Index n20/D 1.470 (literature)
Linear Formula {[(CH3)2 CH]2 N}2 POCH2CH2CN
MDL Number MFCD00012213
Quantity 1 g
Synonym Bis(diisopropylamino)(2-cyanoethoxy)phosphine
Molecular Weight (g/mol) 301.41
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.