Learn More
Strem, An Ascensus Company CAS# 102691-36-1. 1g. 2-Cyanoethyl N,N,N',N'-tetra(i-propyl)phosphorodiamidite, min. 98%. MFCD00012213
SDP
Catalog No. 501385382
Encompass
CAS# 102691-36-1. 1g. 2-Cyanoethyl N,N,N',N'-tetra(i-propyl)phosphorodiamidite, min. 98%. MFCD00012213. Molecular Weight: 301.41. Molecular Formula: {[(CH3)2CH]2N}POCH2CH2CN. Color/form: colorless liq. Strem# 15-0695. http://www.strem.com/catalog/v/15-0695/
Catalog No. 50-138-5382 Supplier Strem, An Ascensus Company Supplier No. 1506951G
CAS# 102691-36-1. 1g. 2-Cyanoethyl N,N,N',N'-tetra(i-propyl)phosphorodiamidite, min. 98%. MFCD00012213. Molecular Weight: 301.41. Molecular Formula: {[(CH3)2CH]2N}POCH2CH2CN. Color/form: colorless liq. Strem# 15-0695. http://www.strem.com/catalog/v/15-0695/