Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanophenylboronic Acid 1,3-Propanediol Ester 97.0+%, TCI America™
SDP

Catalog No. C29935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29935G 5 g
C29931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29935G Supplier TCI America Supplier No. C29935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 172732-52-4
Molecular Formula C10H10BNO2
Molecular Weight (g/mol) 187.005
MDL Number MFCD04039012
InChI Key REQZFVYFYAZUMG-UHFFFAOYSA-N
Synonym 2-1,3,2-dioxaborinan-2-yl benzonitrile, 2-cyanophenylboronic acid 1,3-propanediol ester, 2-2-cyanophenyl-1,3,2-dioxaborinane, 2-cyanobenzeneboronic acid 1,3-propanediol ester, 2-cyanophenylboronic acid propanediol ester, 2-cyanophenylboronic acid, 1,3-propanediol cyclic ester, 2-1,3,2-dioxaborinane-2-yl benzonitrile, 2-1,3,2-dioxaborinan-2-yl-benzonitrile, benzonitrile, 2-1,3,2-dioxaborinan-2-yl, 2-cyanophenylboronic acid,1,3-propanediol
PubChem CID 3529602
IUPAC Name 2-(1,3,2-dioxaborinan-2-yl)benzonitrile
SMILES B1(OCCCO1)C2=CC=CC=C2C#N

Specifications

Melting Point 49°C
Quantity 5 g
Formula Weight 187.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanophenylboronic Acid 1,3-Propanediol Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.