Learn More
Medchemexpress LLC 2-Cyanophenylboronic acid | 138642-62-3 | 99.9% | 146.94 | 10 G
SDP
Catalog No. 5000404090
Encompass_Preferred
2-Cyanophenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It contains both nucleophilic and electrophilic centers. It can function as an efficient electrolyte additive for high voltage LCO cathode.
  • Enhances the structural reversibility
  • Decreases hydrofluoric acid formation
Catalog No. 50-004-04090 Supplier Medchemexpress LLC Supplier No. HY3446910G
2-Cyanophenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It contains both nucleophilic and electrophilic centers. It can function as an efficient electrolyte additive for high voltage LCO cathode.
  • Enhances the structural reversibility
  • Decreases hydrofluoric acid formation