Learn More
Medchemexpress LLC 2-Cyanophenylboronic acid | 138642-62-3 | MFCD01632208 | >99.9% | 146.94 | 5 G
SDP
Catalog No. 5000404458
Encompass_Preferred
2-Cyanophenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It contains both nucleophilic and electrophilic centers. It can function as an efficient electrolyte additive for high voltage LCO cathode, enhancing structural reversibility and decreasing hydrofluoric acid formation.
  • Used as a biological material or organic compound for life science research
  • Contains both nucleophilic and electrophilic centers
  • Functions as an efficient electrolyte additive for high voltage LCO cathode
  • Enhances structural reversibility
  • Decreases hydrofluoric acid formation
Catalog No. 50-004-04458 Supplier Medchemexpress LLC Supplier No. HY344695G
2-Cyanophenylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It contains both nucleophilic and electrophilic centers. It can function as an efficient electrolyte additive for high voltage LCO cathode, enhancing structural reversibility and decreasing hydrofluoric acid formation.
  • Used as a biological material or organic compound for life science research
  • Contains both nucleophilic and electrophilic centers
  • Functions as an efficient electrolyte additive for high voltage LCO cathode
  • Enhances structural reversibility
  • Decreases hydrofluoric acid formation