Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyanopyridine, 98%

Catalog No. AC110870025
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110870025 2.5 kg Plastic bottle
AC110870250 25 g Glass Bottle
AC110871000 100 g Plastic bottle
AC110875000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC110870025 Supplier Thermo Scientific Chemicals Supplier No. 110870025
Only null left
Add to Cart

CAS: 100-48-1 | C6H4N2 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-48-1
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00006417
InChI Key GPHQHTOMRSGBNZ-UHFFFAOYSA-N
Synonym 4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
PubChem CID 7506
ChEBI CHEBI:28020
IUPAC Name pyridine-4-carbonitrile
SMILES C1=CN=CC=C1C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 77°C to 81°C
Color White
Boiling Point 196°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C5H4N)CN
Quantity 2.5 kg
Beilstein 22,46
Solubility Information Solubility in water: 32g/L (16°C)
Formula Weight 104.11
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4-Cyanopyridine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-856-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.