Learn More
Medchemexpress LLC 2-cyclohexen-1-one, 3-(15-hydroxypentadecyl)-2,4,4-trimethyl- | 220757-88-0 | 99.1% | 364.60 | C24H44O2 | 10 MG
SDP
Catalog No. 5000241895
Encompass_Preferred
tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol (C24H44O2, MW 364.60) that promotes neuronal differentiation and modulates Notch1 signaling. It is supplied as a high-purity research standard intended for in vitro studies of neural stem cell differentiation and neuronal signaling pathways.
  • Promotes differentiation of neural stem cell-derived neurospheres toward neurons.
  • Modulates Notch1 expression and signaling.
  • High purity suitable for in vitro research applications.
  • Soluble in DMSO for solution-based assays.
  • Shown to stimulate arginine vasopressin secretion in neurohypophysis preparations.
Catalog No. 50-002-41895 Supplier Medchemexpress LLC Supplier No. HY10403110MG
tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol (C24H44O2, MW 364.60) that promotes neuronal differentiation and modulates Notch1 signaling. It is supplied as a high-purity research standard intended for in vitro studies of neural stem cell differentiation and neuronal signaling pathways.
  • Promotes differentiation of neural stem cell-derived neurospheres toward neurons.
  • Modulates Notch1 expression and signaling.
  • High purity suitable for in vitro research applications.
  • Soluble in DMSO for solution-based assays.
  • Shown to stimulate arginine vasopressin secretion in neurohypophysis preparations.