Learn More
Medchemexpress LLC 2-cyclohexen-1-one, 3-(15-hydroxypentadecyl)-2,4,4-trimethyl- | 220757-88-0 | 99.1% | 364.60 | C24H44O2 | 5 MG
SDP
Catalog No. 5000243169
Encompass_Preferred
tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol reported to promote neuronal differentiation and modulate Notch signaling. It is supplied as a pure-form research reagent with molecular formula C24H44O2 and molecular weight 364.60, and has documented analytical purity and storage stability.
  • Promotes neuronal differentiation and modulates Notch signaling.
  • High purity (99.12% by LCMS).
  • Long-chain fatty alcohol structure with a 15-carbon side chain.
  • Stable in pure form for extended storage at -20°C.
  • Intended for research use in neuronal and stem cell pathway studies.
Catalog No. 50-002-43169 Supplier Medchemexpress LLC Supplier No. HY1040315MG
tCFA15 is a trimethyl cyclohexenonic long-chain fatty alcohol reported to promote neuronal differentiation and modulate Notch signaling. It is supplied as a pure-form research reagent with molecular formula C24H44O2 and molecular weight 364.60, and has documented analytical purity and storage stability.
  • Promotes neuronal differentiation and modulates Notch signaling.
  • High purity (99.12% by LCMS).
  • Long-chain fatty alcohol structure with a 15-carbon side chain.
  • Stable in pure form for extended storage at -20°C.
  • Intended for research use in neuronal and stem cell pathway studies.