Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Cyclohexyl-4-methylphenol | 1596-09-4 | 99.6% | C13H18O | 100 MG
SDP

Catalog No. 5000433597
Encompass_Preferred

2-Cyclohexyl-4-methylphenol, also known as 4-Methyl-2-cyclohexylphenol, is classified as a drug intermediate. It is utilized in the synthesis of various active compounds. This substance has a molecular weight of 190.29 and appears as a light yellow to yellow oil. It is intended for research use only.

  • Intermediate for the synthesis of active compounds.
  • Molecular Weight: 190.29.
  • Appearance: Oil.
  • Color: Light yellow to yellow.
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years.
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month.
  • Shipping: Room temperature in continental US.

Catalog No. 50-004-33597 Supplier Medchemexpress LLC Supplier No. HY75323100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Cyclohexyl-4-methylphenol, also known as 4-Methyl-2-cyclohexylphenol, is classified as a drug intermediate. It is utilized in the synthesis of various active compounds. This substance has a molecular weight of 190.29 and appears as a light yellow to yellow oil. It is intended for research use only.

  • Intermediate for the synthesis of active compounds.
  • Molecular Weight: 190.29.
  • Appearance: Oil.
  • Color: Light yellow to yellow.
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years.
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month.
  • Shipping: Room temperature in continental US.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.