Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclohexylcyclohexanone 97.0+%, TCI America™
SDP

Catalog No. C06125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06125G 5 g
C061225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06125G Supplier TCI America Supplier No. C06125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90-42-6
Molecular Formula C12H20O
Molecular Weight (g/mol) 180.29
MDL Number MFCD00013795
InChI Key UOBQDYFTAJKQAL-UHFFFAOYNA-N
PubChem CID 7016
IUPAC Name [1,1'-bi(cyclohexane)]-2-one
SMILES O=C1CCCCC1C1CCCCC1

Specifications

Color Yellow
Boiling Point 269°C
Quantity 5 g
Formula Weight 180.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexylcyclohexanone
RTECSNumber DT7400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.