Learn More
Cayman Chemical 2-(cyclohexylmethyl)-Plumbagin, Purity: ≥95%, CAS: 3033734-59-4, 10 mg
SDP
Catalog No. 502918393
Encompass_Preferred
A derivative of plumbagin; selectively cytotoxic to PANC-1 cells in nutrient-deprived medium over PANC-1 cells in DMEM (PC50s = 0.11 and 47.2 µM, respectively); induces apoptosis in PANC-1 cells at 1 µM; selectively reduces Akt and mTOR phosphorylation in PANC-1 cells under nutrient-deprived, but not nutrient-rich, conditions; tumor volume and weight in a MiaPaCa-2 mouse xenograft model at 50 and 250 µg/animal five times per week
Catalog No. 50-291-8393 Supplier Cayman Chemical Supplier No. 3914310 MG
A derivative of plumbagin; selectively cytotoxic to PANC-1 cells in nutrient-deprived medium over PANC-1 cells in DMEM (PC50s = 0.11 and 47.2 µM, respectively); induces apoptosis in PANC-1 cells at 1 µM; selectively reduces Akt and mTOR phosphorylation in PANC-1 cells under nutrient-deprived, but not nutrient-rich, conditions; tumor volume and weight in a MiaPaCa-2 mouse xenograft model at 50 and 250 µg/animal five times per week