Learn More
Medchemexpress LLC 2-Cyclopenten-1-one, 3-methyl- | 2758-18-1 | 99.12% | 96.13 | 10 G
SDP
Catalog No. 5000451459
Encompass_Preferred
3-Methyl-2-cyclopenten-1-one is an endogenous metabolite provided for research use. It is a light yellow to brown liquid with a molecular weight of 96.13.
  • Intended for research and analytical use.
  • Available in various quantities including 10 gram.
  • Stored at -20°C for 3 years or 4°C for 2 years in pure form.
  • Stored at -80°C for 6 months or -20°C for 1 month in solvent.
  • Appearance is a light yellow to brown liquid.
Catalog No. 50-004-51459 Supplier Medchemexpress LLC Supplier No. HYW01301410G
3-Methyl-2-cyclopenten-1-one is an endogenous metabolite provided for research use. It is a light yellow to brown liquid with a molecular weight of 96.13.
  • Intended for research and analytical use.
  • Available in various quantities including 10 gram.
  • Stored at -20°C for 3 years or 4°C for 2 years in pure form.
  • Stored at -80°C for 6 months or -20°C for 1 month in solvent.
  • Appearance is a light yellow to brown liquid.