Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Cyclopenten-1-one

Catalog No. C1129095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1129095G 5 g
C11290925G 25 g
C112909100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1129095G Supplier Sigma Aldrich Supplier No. C1129095G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Versatile electrophile employed in a variety of addition reactions including conjugate addition of organocopper nucleophiles, Michael reaction with silyl enol ethers, and siloxanes, Diels-Alder cycloadditions, and phosphoniosilylations.

Specifications

CAS 930-30-3
Density 0.98 g/mL (at 25°C (literature))
Boiling Point 64°C to 65°C (19 mmHg)
Molecular Formula C5H6O
Linear Formula C5H6O
Refractive Index n20/D 1.481 (literature)
MDL Number MFCD00001401
Quantity 5 g
Molecular Weight (g/mol) 82.1
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.